siRNA Details
Gene Symbol: FRG2KP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135055.1 | 4 | 563 | |||
GenBank mRNA | BI521010.1 | - | 597 |
Gene Symbol: LOC107983990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255752.3 | XP_005255809.1 | 4 | 648 |
