siRNA Details
| Gene Symbol: LOC440390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126008.1 | 1 | 868 | |||
| GenBank mRNA | AK128193.1 | - | 1 | 868 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC440390| Gene Symbol: LOC440390 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126008.1 | 1 | 868 | |||
| GenBank mRNA | AK128193.1 | - | 1 | 868 | ||