siRNA Details
| Gene Symbol: CCDC144NL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC072410.1 | - | 3 | 1224 | ||
| BC113722.1 | - | 1 | 806 | |||
| BC117395.1 | - | 1 | 806 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC144NL-AS1| Gene Symbol: CCDC144NL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC072410.1 | - | 3 | 1224 | ||
| BC113722.1 | - | 1 | 806 | |||
| BC117395.1 | - | 1 | 806 | |||