siRNA Details
| Gene Symbol: LRRC37A8P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126038.1 | 1 | 1371 | |||
| GenBank mRNA | AK125932.1 | - | 1 | 297 | ||
| AK128235.1 | - | 1 | 1371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LRRC37A8P| Gene Symbol: LRRC37A8P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126038.1 | 1 | 1371 | |||
| GenBank mRNA | AK125932.1 | - | 1 | 297 | ||
| AK128235.1 | - | 1 | 1371 | |||