siRNA Details
| Gene Symbol: EMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018447.3 | NP_060917.1 | 4 | 757 | ||
| XM_005265321.3 | XP_005265378.1 | 4 | 625 | |||
| GenBank mRNA | AF157321.1 | - | 3 | 469 | ||
| AK172843.1 | - | 3 | 465 | |||
| AK222953.1 | - | 3 | 467 | |||
| AK312008.1 | - | 3 | 464 | |||
| BC022807.1 | - | 3 | 452 | 4897577 | ||

EMC3
Genomic Map