siRNA Details
| Gene Symbol: WFDC21P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030732.1 | 3 | 446 | |||
| GenBank mRNA | AF086204.1 | - | 3 | 272 | ||
| KJ020271.1 | - | 3 | 243 | |||
| KJ020272.1 | - | 2 | 223 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WFDC21P| Gene Symbol: WFDC21P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030732.1 | 3 | 446 | |||
| GenBank mRNA | AF086204.1 | - | 3 | 272 | ||
| KJ020271.1 | - | 3 | 243 | |||
| KJ020272.1 | - | 2 | 223 | |||