siRNA Details
| Gene Symbol: HSBP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136180.1 | NP_001129652.1 | 3 | 306 | ||
| GenBank mRNA | AK001829.1 | - | 1 | 630 | ||
| BC017654.1 | - | 3 | 234 | |||
| BC040056.2 | - | 3 | 230 | |||
| BC157848.1 | - | 3 | 228 | 9057067 | ||

HSBP1L1
Genomic Map