siRNA Details
| Gene Symbol: NBPF13P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103466.1 | 5 | 545 | |||
| GenBank mRNA | BX336604.2 | - | 5 | 545 | ||
| CB154168.1 | - | 3 | 222 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NBPF13P| Gene Symbol: NBPF13P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103466.1 | 5 | 545 | |||
| GenBank mRNA | BX336604.2 | - | 5 | 545 | ||
| CB154168.1 | - | 3 | 222 | |||