siRNA Details
Gene Symbol: LOC284561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK001966.1 | - | 2 | 1017 | ||
AK023548.1 | - | 4 | 761 | |||
AK023809.1 | - | 2 | 997 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC284561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK001966.1 | - | 2 | 1017 | ||
AK023548.1 | - | 4 | 761 | |||
AK023809.1 | - | 2 | 997 |