siRNA Details
| Gene Symbol: RASAL2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027982.1 | 2 | 601 | |||
| GenBank mRNA | AK124464.1 | - | 2 | 235 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RASAL2-AS1| Gene Symbol: RASAL2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027982.1 | 2 | 601 | |||
| GenBank mRNA | AK124464.1 | - | 2 | 235 | ||