siRNA Details
| Gene Symbol: ZNF337-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126467.1 | 2 | 218 | |||
| GenBank mRNA | AL529206.3 | - | 2 | 218 | ||
| BC034569.1 | - | 2 | 107 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF337-AS1| Gene Symbol: ZNF337-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126467.1 | 2 | 218 | |||
| GenBank mRNA | AL529206.3 | - | 2 | 218 | ||
| BC034569.1 | - | 2 | 107 | |||