siRNA Details
| Gene Symbol: LINC01272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278655.1 | NP_001265584.1 | 4 | 1544 | ||
| XM_006723680.3 | XP_006723743.1 | 4 | 1557 | |||
| GenBank mRNA | BI861769.1 | - | 1 | 759 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01272| Gene Symbol: LINC01272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278655.1 | NP_001265584.1 | 4 | 1544 | ||
| XM_006723680.3 | XP_006723743.1 | 4 | 1557 | |||
| GenBank mRNA | BI861769.1 | - | 1 | 759 | ||