siRNA Details
| Gene Symbol: LINC01272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278655.1 | NP_001265584.1 | 4 | 535 | ||
| XM_006723680.3 | XP_006723743.1 | 4 | 548 | |||
| GenBank mRNA | AK307192.1 | - | 4 | 523 | ||
| BQ430993.1 | - | 4 | 535 | |||

LINC01272
Genomic Map