siRNA Details
| Gene Symbol: MIAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003491.3 | 3 | 285 | |||
| NR_033319.2 | 3 | 285 | ||||
| GenBank mRNA | AB263414.1 | - | 3 | 247 | ||
| AB263415.1 | - | 3 | 247 | |||
| AK098833.1 | - | 3 | 179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIAT| Gene Symbol: MIAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003491.3 | 3 | 285 | |||
| NR_033319.2 | 3 | 285 | ||||
| GenBank mRNA | AB263414.1 | - | 3 | 247 | ||
| AB263415.1 | - | 3 | 247 | |||
| AK098833.1 | - | 3 | 179 | |||