siRNA Details
| Gene Symbol: MIAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003491.3 | 5 | 9944 | |||
| NR_033319.2 | 4 | 9818 | ||||
| NR_033320.2 | 5 | 9870 | ||||
| NR_033321.2 | 4 | 9744 | ||||
| GenBank mRNA | AB263414.1 | - | 5 | 9905 | ||
| AB263415.1 | - | 4 | 9779 | |||
| AB263416.1 | - | 5 | 9831 | |||
| AB263417.1 | - | 4 | 9705 | |||
| AK098833.1 | - | 4 | 1476 | |||
| AK127256.1 | - | 1 | 5056 | |||

MIAT
Genomic Map