siRNA Details
| Gene Symbol: LOC440934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_136638.1 | 2 | 957 | |||
| NR_136639.1 | 2 | 661 | ||||
| NR_136640.1 | 2 | 226 | ||||
| NR_136641.1 | 2 | 405 | ||||
| GenBank mRNA | AA401699.1 | - | 2 | 212 | ||
| BC008048.2 | - | 2 | 956 | |||
| BC033986.1 | - | 2 | 627 | |||
| HY021245.1 | - | 2 | 227 | |||

LOC440934
Genomic Map