siRNA Details
| Gene Symbol: TMEM106B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134232.1 | NP_001127704.1 | 5 | 748 | ||
| NM_018374.3 | NP_060844.2 | 6 | 942 | |||
| XM_005249789.1 | XP_005249846.1 | 5 | 798 | |||
| GenBank mRNA | AK002135.1 | - | 5 | 672 | ||
| AK223263.1 | - | 5 | 646 | |||
| AK311651.1 | - | 6 | 901 | |||
| BC028108.1 | - | 5 | 662 | |||
| BC033901.2 | - | 6 | 864 | 5263860 | ||
| BC039741.2 | - | 5 | 659 | 6066301 | ||

TMEM106B
Genomic Map