siRNA Details
Gene Symbol: ZNF654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017006789.1 | XP_016862278.1 | 4, 5 | 642 | ||
XM_017006790.1 | XP_016862279.1 | 4, 5 | 654 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF654 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017006789.1 | XP_016862278.1 | 4, 5 | 642 | ||
XM_017006790.1 | XP_016862279.1 | 4, 5 | 654 |