siRNA Details
| Gene Symbol: ZNF654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011534340.2 | XP_011532642.2 | 4 | 636 | ||
| XM_017006789.1 | XP_016862278.1 | 5 | 765 | |||
| XM_017006790.1 | XP_016862279.1 | 5 | 777 | |||
| XM_017006793.1 | XP_016862282.1 | 4 | 636 | |||
| XM_017006794.1 | XP_016862283.1 | 3 | 376 | |||

ZNF654
Genomic Map