siRNA Details
| Gene Symbol: LINC00613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103762.1 | 3, 4 | 298 | |||
| NR_103763.1 | 2, 3 | 138 | ||||
| GenBank mRNA | AW237894.1 | - | 3, 4 | 319 | ||
| BG204498.1 | - | 1, 2 | 69 | |||
| BG220241.1 | - | 2, 3 | 138 | |||

LINC00613
Genomic Map