siRNA Details
| Gene Symbol: C5orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023073.3 | NP_075561.3 | 10 | 1452 | ||
| XM_005248345.3 | XP_005248402.1 | 10 | 1440 | |||
| XM_005248346.3 | XP_005248403.1 | 10 | 1360 | |||
| XM_005248347.3 | XP_005248404.1 | 10 | 1360 | |||
| XM_005248349.3 | XP_005248406.1 | 10 | 1360 | |||
| XM_005248350.3 | XP_005248407.1 | 10 | 1358 | |||
| XM_011514085.2 | XP_011512387.1 | 10 | 1359 | |||
| XM_011514086.2 | XP_011512388.1 | 10 | 1356 | |||
| XM_011514087.2 | XP_011512389.1 | 10 | 1445 | |||
| XM_011514088.2 | XP_011512390.1 | 10 | 1445 | |||
| XM_011514089.2 | XP_011512391.1 | 10 | 1445 | |||
| XM_011514090.2 | XP_011512392.1 | 10 | 1223 | |||
| XM_011514092.2 | XP_011512394.1 | 10 | 1445 | |||
| XM_017009760.1 | XP_016865249.1 | 9 | 1171 | |||
| XM_017009761.1 | XP_016865250.1 | 9 | 1139 | |||
| XM_017009762.1 | XP_016865251.1 | 6 | 898 | |||
| XM_017009763.1 | XP_016865252.1 | 6 | 771 | |||
| XM_017009764.1 | XP_016865253.1 | 6 | 732 | |||
| XM_017009765.1 | XP_016865254.1 | 10 | 1140 | |||

C5orf42
Genomic Map