siRNA Details
| Gene Symbol: C5orf67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287053.1 | NP_001273982.1 | 2 | 242 | ||
| XM_017008942.1 | XP_016864431.1 | 4 | 1362 | |||
| GenBank mRNA | BX089473.1 | - | 2 | 240 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C5orf67| Gene Symbol: C5orf67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287053.1 | NP_001273982.1 | 2 | 242 | ||
| XM_017008942.1 | XP_016864431.1 | 4 | 1362 | |||
| GenBank mRNA | BX089473.1 | - | 2 | 240 | ||