siRNA Details
| Gene Symbol: CHST11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173982.1 | NP_001167453.1 | 3 | 1343 | ||
| NM_018413.5 | NP_060883.1 | 3 | 1358 | |||
| XM_017019369.1 | XP_016874858.1 | 5 | 929 | |||
| GenBank mRNA | AB042326.1 | - | 3 | 1246 | ||
| AF239820.1 | - | 3 | 1009 | |||
| AJ289134.1 | - | 3 | 985 | |||
| AK290923.1 | - | 3 | 1299 | |||
| AK313217.1 | - | 3 | 1286 | |||
| BC013315.1 | - | 4 | 1155 | 4042813 | ||

CHST11
Genomic Map