siRNA Details
| Gene Symbol: MEIKIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303622.1 | NP_001290551.1 | 8 | 877 | ||
| GenBank mRNA | AB987829.1 | - | 8 | 684 | ||
| DY654845.1 | - | 4 | 276 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MEIKIN| Gene Symbol: MEIKIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303622.1 | NP_001290551.1 | 8 | 877 | ||
| GenBank mRNA | AB987829.1 | - | 8 | 684 | ||
| DY654845.1 | - | 4 | 276 | |||