siRNA Details
| Gene Symbol: LYRM4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126016.1 | 3 | 486 | |||
| NR_126017.1 | 3 | 404 | ||||
| NR_126018.1 | 2 | 391 | ||||
| NR_126019.1 | 2 | 473 | ||||
| GenBank mRNA | BC042032.1 | - | 2 | 391 | ||
| BC133670.1 | - | 2 | 414 | |||
| BE788607.1 | - | 3 | 319 | |||
| CR736828.1 | - | 3 | 456 | |||

LYRM4-AS1
Genomic Map