siRNA Details
| Gene Symbol: TAF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138572.2 | NP_612639.2 | 9 | 2177 | ||
| XM_011514296.1 | XP_011512598.1 | 10 | 2290 | |||
| XM_017010243.1 | XP_016865732.1 | 9 | 2331 | |||
| XM_017010245.1 | XP_016865734.1 | 7 | 1875 | |||
| GenBank mRNA | AF086309.1 | - | 1 | 420 | ||
| AL833401.1 | - | 10 | 2289 | |||

TAF8
Genomic Map