siRNA Details
| Gene Symbol: OOEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080507.2 | NP_001073976.1 | 2 | 206 | ||
| XM_017010890.1 | XP_016866379.1 | 2 | 185 | |||
| GenBank mRNA | BX093821.1 | - | 2 | 196 | ||
| EU290647.1 | - | 2 | 206 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OOEP| Gene Symbol: OOEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080507.2 | NP_001073976.1 | 2 | 206 | ||
| XM_017010890.1 | XP_016866379.1 | 2 | 185 | |||
| GenBank mRNA | BX093821.1 | - | 2 | 196 | ||
| EU290647.1 | - | 2 | 206 | |||