siRNA Details
| Gene Symbol: EMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018447.3 | NP_060917.1 | 9 | 1237 | ||
| GenBank mRNA | AF157321.1 | - | 8 | 949 | ||
| AK172843.1 | - | 8 | 945 | |||
| AK312008.1 | - | 8 | 944 | |||
| BC022807.1 | - | 8 | 932 | 4897577 | ||

EMC3
Genomic Map