siRNA Details
| Gene Symbol: CAND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018448.4 | NP_060918.2 | 10 | 3187 | ||
| GenBank mRNA | AB020636.2 | - | 10 | 3089 | ||
| AK001791.1 | - | 1 | 588 | |||
| AK027404.1 | - | 4 | 1822 | |||
| AK074922.1 | - | 7 | 2361 | |||
| AK292456.1 | - | 12 | 3026 | |||
| AK314358.1 | - | 10 | 3075 | |||
| AL136810.1 | - | 10 | 3099 | |||
| AL833880.2 | - | 4 | 1370 | |||
| BC004232.1 | - | 1 | 740 | |||
| BC026220.2 | - | 8 | 2538 | |||
| BC050341.1 | - | 10 | 2883 | 5265409 | ||

CAND1
Genomic Map