siRNA Details
| Gene Symbol: TMEM242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018452.5 | NP_060922.2 | 4 | 3474 | ||
| GenBank mRNA | BC029130.1 | - | 4 | 2736 | 5182086 | |
| CD244433.1 | - | 1 | 141 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM242| Gene Symbol: TMEM242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018452.5 | NP_060922.2 | 4 | 3474 | ||
| GenBank mRNA | BC029130.1 | - | 4 | 2736 | 5182086 | |
| CD244433.1 | - | 1 | 141 | |||