siRNA Details
| Gene Symbol: LOC101929709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125822.1 | 1 | 646 | |||
| GenBank mRNA | BG497825.1 | - | 1 | 242 | ||
| BQ652687.1 | - | 1 | 229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929709| Gene Symbol: LOC101929709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125822.1 | 1 | 646 | |||
| GenBank mRNA | BG497825.1 | - | 1 | 242 | ||
| BQ652687.1 | - | 1 | 229 | |||