siRNA Details
| Gene Symbol: LOC105376256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK130710.1 | - | 2 | 604 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105376256| Gene Symbol: LOC105376256 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK130710.1 | - | 2 | 604 | ||