siRNA Details
| Gene Symbol: TMEM176A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018487.2 | NP_060957.2 | 7 | 767 | ||
| XM_011516376.2 | XP_011514678.1 | 7 | 902 | |||
| XM_011516377.2 | XP_011514679.1 | 7 | 918 | |||
| XM_011516378.2 | XP_011514680.1 | 6 | 800 | |||
| XM_017012393.1 | XP_016867882.1 | 7 | 1003 | |||
| GenBank mRNA | AF258340.1 | - | 7 | 742 | ||
| AK123468.1 | - | 2 | 1793 | |||
| BC008303.1 | - | 7 | 785 | 3536986 | ||

TMEM176A
Genomic Map