siRNA Details
| Gene Symbol: ZFAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003604.2 | 2 | 610 | |||
| NR_003605.1 | 2 | 291 | ||||
| NR_003606.2 | 2 | 610 | ||||
| NR_036658.1 | 2 | 610 | ||||
| NR_036659.1 | 2 | 168 | ||||
| GenBank mRNA | AY339614.1 | - | 2 | 112 | ||
| AY513722.1 | - | 2 | 610 | |||
| AY513723.1 | - | 2 | 291 | |||
| BC105282.1 | - | 2 | 150 | |||
| BM745942.1 | - | 2 | 170 | |||
| CB137861.1 | - | 2 | 146 | |||
| CR936805.1 | - | 2 | 148 | |||

ZFAS1
Genomic Map