siRNA Details
| Gene Symbol: ZFAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003604.2 | 2 | 635 | |||
| NR_003605.1 | 2 | 316 | ||||
| NR_036658.1 | 2 | 635 | ||||
| NR_036659.1 | 2 | 193 | ||||
| GenBank mRNA | AY339614.1 | - | 2 | 137 | ||
| AY513722.1 | - | 2 | 635 | |||
| AY513723.1 | - | 2 | 316 | |||
| BC105282.1 | - | 2 | 175 | |||
| BM745942.1 | - | 2 | 195 | |||
| CR936805.1 | - | 2 | 173 | |||

ZFAS1
Genomic Map