siRNA Details
| Gene Symbol: PARL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037639.2 | NP_001032728.1 | 2 | 243 | ||
| NM_001324436.1 | NP_001311365.1 | 2 | 243 | |||
| NM_001324437.1 | NP_001311366.1 | 2 | 243 | |||
| NM_001324438.1 | NP_001311367.1 | 2 | 243 | |||
| NM_018622.6 | NP_061092.3 | 2 | 243 | |||
| NR_136893.1 | 2 | 243 | ||||
| XM_005247582.4 | XP_005247639.1 | 2 | 238 | |||
| XM_017006800.1 | XP_016862289.1 | 2 | 238 | |||
| XM_017006801.1 | XP_016862290.1 | 2 | 236 | |||
| XM_017006802.1 | XP_016862291.1 | 2 | 236 | |||
| XM_017006803.1 | XP_016862292.1 | 2 | 234 | |||
| GenBank mRNA | AF197937.1 | - | 2 | 181 | ||
| AK024765.1 | - | 2 | 215 | |||
| BC003653.1 | - | 2 | 230 | 3537438 | ||
| BC014058.2 | - | 2 | 201 | 3937238 | ||
| BG714761.1 | - | 2 | 241 | |||
| CB122278.1 | - | 2 | 240 | |||

PARL
Genomic Map