siRNA Details
| Gene Symbol: ADAM3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001569.3 | 11 | 1310 | |||
| NR_024106.2 | 11 | 1310 | ||||
| NR_024107.2 | 11 | 1310 | ||||
| NR_073423.1 | 11 | 1310 | ||||
| GenBank mRNA | AK302269.1 | - | 8 | 857 | ||
| AK308250.1 | - | 11 | 1174 | |||

ADAM3A
Genomic Map