siRNA Details
| Gene Symbol: RHOXF1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131238.1 | 2 | 547 | |||
| GenBank mRNA | AK123976.1 | - | 2 | 547 | ||
| DA929866.1 | - | 2 | 547 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RHOXF1-AS1| Gene Symbol: RHOXF1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131238.1 | 2 | 547 | |||
| GenBank mRNA | AK123976.1 | - | 2 | 547 | ||
| DA929866.1 | - | 2 | 547 | |||