siRNA Details
| Gene Symbol: DDX43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018665.2 | NP_061135.2 | 1 | 451 | ||
| XM_011535926.1 | XP_011534228.1 | 1 | 451 | |||
| XM_011535928.2 | XP_011534230.1 | 1 | 153 | |||
| XM_017011003.1 | XP_016866492.1 | 1 | 119 | |||
| GenBank mRNA | AJ278110.1 | - | 1 | 314 | ||
| AK303324.1 | - | 1 | 160 | |||
| AK313686.1 | - | 1 | 151 | |||
| AL136751.1 | - | 1 | 139 | |||
| BC066938.1 | - | 1 | 132 | 4829072 | ||
| BX646425.1 | - | 1 | 198 | |||

DDX43
Genomic Map