siRNA Details
| Gene Symbol: LOC107986705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012880.1 | XP_016868369.1 | 6 | 1975 | ||
| XM_017012881.1 | XP_016868370.1 | 6 | 1433 | |||
| XM_017012882.1 | XP_016868371.1 | 4 | 1472 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC107986705| Gene Symbol: LOC107986705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012880.1 | XP_016868369.1 | 6 | 1975 | ||
| XM_017012881.1 | XP_016868370.1 | 6 | 1433 | |||
| XM_017012882.1 | XP_016868371.1 | 4 | 1472 | |||