siRNA Details
| Gene Symbol: GRIFIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291784.1 | NP_001278713.1 | 1, 2 | 95 | ||
| XM_011515405.2 | XP_011513707.1 | 2, 3 | 275 | |||
| GenBank mRNA | CN281639.1 | - | 2, 3 | 254 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRIFIN| Gene Symbol: GRIFIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291784.1 | NP_001278713.1 | 1, 2 | 95 | ||
| XM_011515405.2 | XP_011513707.1 | 2, 3 | 275 | |||
| GenBank mRNA | CN281639.1 | - | 2, 3 | 254 | ||