siRNA Details
| Gene Symbol: RBM27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018989.1 | NP_061862.1 | 2 | 313 | ||
| XM_005268466.1 | XP_005268523.1 | 2 | 363 | |||
| XM_005268467.1 | XP_005268524.1 | 2 | 363 | |||
| XM_005268468.1 | XP_005268525.1 | 2 | 363 | |||
| XM_006714787.1 | XP_006714850.1 | 2 | 363 | |||
| XM_011537661.1 | XP_011535963.1 | 2 | 363 | |||
| XM_011537662.1 | XP_011535964.1 | 2 | 363 | |||
| XM_017009587.1 | XP_016865076.1 | 2 | 363 | |||
| GenBank mRNA | AK128738.1 | - | 2 | 363 | ||
| BC009537.1 | - | 2 | 316 | |||
| BX647384.1 | - | 2 | 313 | |||

RBM27
Genomic Map