siRNA Details
Gene Symbol: LINC00917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024406.1 | 5 | 1648 | |||
GenBank mRNA | AK093244.1 | - | 5 | 1648 | ||
BC043250.1 | - | 4 | 1705 | |||
BC144028.1 | - | 3 | 868 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024406.1 | 5 | 1648 | |||
GenBank mRNA | AK093244.1 | - | 5 | 1648 | ||
BC043250.1 | - | 4 | 1705 | |||
BC144028.1 | - | 3 | 868 |