siRNA Details
Gene Symbol: ZNF823 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080493.3 | NP_001073962.1 | 1 | 151 | ||
NM_001297610.1 | NP_001284539.1 | 1 | 151 | |||
NM_017507.1 | NP_059977.1 | 1 | 151 | |||
XM_017026941.1 | XP_016882430.1 | 1 | 139 | |||
XM_017026942.1 | XP_016882431.1 | 1 | 139 | |||
GenBank mRNA | AK300579.1 | - | 1 | 118 | ||
AK303213.1 | - | 1 | 116 | |||
AK308189.1 | - | 1 | 87 | |||
BC052597.1 | - | 1 | 63 | 4304302 | ||
DA519419.1 | - | 1 | 151 | |||
DA880101.1 | - | 1 | 87 | |||
X51760.1 | - | 1 | 65 |
