siRNA Details
| Gene Symbol: ZNF823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080493.3 | NP_001073962.1 | 4 | 2437 | ||
| NM_001297610.1 | NP_001284539.1 | 3 | 2376 | |||
| NM_017507.1 | NP_059977.1 | 3 | 2310 | |||
| XM_017026941.1 | XP_016882430.1 | 4 | 2599 | |||
| XM_017026942.1 | XP_016882431.1 | 2 | 2237 | |||
| GenBank mRNA | AK303213.1 | - | 3 | 2341 | ||
| BC052597.1 | - | 4 | 2349 | 4304302 | ||
| BC063560.1 | - | 1 | 1610 | |||

ZNF823
Genomic Map