siRNA Details
| Gene Symbol: YLPM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019589.2 | NP_062535.2 | 3 | 1371 | ||
| XM_005267860.3 | XP_005267917.1 | 3 | 1415 | |||
| XM_011536966.2 | XP_011535268.1 | 4 | 530 | |||
| XM_011536967.1 | XP_011535269.1 | 3 | 1415 | |||
| GenBank mRNA | AK090435.1 | - | 1 | 127 | ||
| AK095760.1 | - | 3 | 1371 | |||
| BC044636.1 | - | 3 | 732 | |||
| DB096717.1 | - | 2 | 357 | |||

YLPM1
Genomic Map