siRNA Details
| Gene Symbol: YLPM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019589.2 | NP_062535.2 | 10 | 5461 | ||
| XM_005267860.3 | XP_005267917.1 | 10 | 5505 | |||
| XM_011536966.2 | XP_011535268.1 | 11 | 4620 | |||
| XM_011536967.1 | XP_011535269.1 | 9 | 3387 | |||
| GenBank mRNA | AK090435.1 | - | 8 | 4217 | ||
| AK297582.1 | - | 6 | 1032 | |||
| AL832365.1 | - | 7 | 3996 | |||
| BC007792.1 | - | 1 | 58 | |||
| L40403.1 | - | 4 | 564 | |||

YLPM1
Genomic Map