siRNA Details
| Gene Symbol: ZNF630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037735.3 | NP_001032824.2 | 5 | 894 | ||
| NM_001190255.2 | NP_001177184.1 | 5 | 968 | |||
| NM_001282201.1 | NP_001269130.1 | 5 | 1209 | |||
| NM_001282202.1 | NP_001269131.1 | 5 | 1205 | |||
| GenBank mRNA | AK000580.1 | - | 3 | 625 | ||
| AK307979.1 | - | 5 | 869 | |||
| BC037316.1 | - | 5 | 979 | |||
| BC112139.1 | - | 4 | 707 | 8327607 | ||

ZNF630
Genomic Map