siRNA Details
| Gene Symbol: PPM1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020700.1 | NP_065751.1 | 10 | 1783 | ||
| XM_011538578.2 | XP_011536880.1 | 10 | 1919 | |||
| XM_011538579.1 | XP_011536881.1 | 10 | 1471 | |||
| GenBank mRNA | AB032983.1 | - | 9 | 1376 | ||
| AB084258.1 | - | 10 | 2048 | |||
| BC157843.1 | - | 10 | 1793 | 9057062 | ||

PPM1H
Genomic Map