siRNA Details
| Gene Symbol: KIAA1211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020722.1 | NP_065773.1 | 11 | 5161 | ||
| XM_005265752.2 | XP_005265809.1 | 10 | 5168 | |||
| XM_005265755.2 | XP_005265812.1 | 7 | 4597 | |||
| XM_005265756.2 | XP_005265813.1 | 7 | 4645 | |||
| XM_006714045.2 | XP_006714108.1 | 7 | 4622 | |||
| XM_011534397.2 | XP_011532699.1 | 10 | 5911 | |||
| XM_017008473.1 | XP_016863962.1 | 10 | 4897 | |||
| XM_017008474.1 | XP_016863963.1 | 9 | 5775 | |||
| GenBank mRNA | AB033037.1 | - | 5 | 3482 | ||
| AL133028.1 | - | 4 | 3573 | |||

KIAA1211
Genomic Map